ContactUsLinkComponent
VWR® Pipette Carousel Stand
VWR® Pipette Carousel Stand
Catalog #: 10827-940
Supplier:  VWR International
VWR* Pipette stand 7-place
VWR® Pipette Carousel Stand
Catalog #: 10827-940
Supplier:  VWR International
Specifications
  • Description:
    VWR® Carousel Pipettor Stand
  • Cat. no.:
    10827-940
Specifications
About this item
Rotary pipette stand for up 7 pipettors. Made from PP/PC.
  • Pipettes hang vertically to prevent liquids from running inside the shaft
  • Rotating head for easy access
  • Designed for single and multichannel models
Customers Who Bought This Also Bought