Corepoint® Scientific Hazardous Location Manual Defrost Freezers
Supplier: Horizon Scientific
Certificates
About this item
These freezers are designed to keep you and your stored substances safe in work areas where hazardous conditions may exist.
- Laboratory use
- Microprocessor temperature controller
- Temperature alarms
- Keyed door locks
- Natural hydrocarbon refrigerants
- ENERGY STAR certified*
These Hazardous Location Laboratory Freezers meet NFPA and OSHA guidelines for Hazardous Location, Class 1, Division II, Groups C and D guidelines. These units utilize non-sparking and/or ATEX rated components and are engineered for use in environments where volatile/explosive conditions could potentially exist.
Models feature a microprocessor temperature controller, allowing for precise temperature control, recovery and maintenance versus mechanical style thermostats. For storage of flammable or volatile materials within typical laboratory or non-volatile environments, see flammable material storage units.
Certifications: ETL listed
*Select models ENERGY STAR certified: 76717-348, 76717-350, and 76717-352
*Select models ENERGY STAR certified: 76717-348, 76717-350, and 76717-352
Specifications
- Catalog No:
- 76717-352
- 76717-350
- 76717-348
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Cat. No.:
- 76717-352
- 76717-350
- 76717-348
- Defrost:
- Manual
- Manual
- Manual
- Shelves/drawers:
- 6 Fixed shelves
- 6 Fixed shelves
- 6 Fixed shelves
- Door type:
- 1 Solid door, right hinged
- 1 Solid door, right hinged
- 1 Solid door, right hinged
- Weight:
- 102 kg (225 lbs.)
- 98 kg (215 lbs.)
- 77 kg (170 lbs.)
Specifications
Related Information
TDS 14 cu. ft. Hazardous Location Manual Defrost Freezer TDS 17 cu. ft. Hazardous Location Manual Defrost Freezer TDS 20 cu. ft. Hazardous Location Manual Defrost Freezer TDS Corepoint® Scientific Hazardous Location Manual Defrost Freezers, 14 cu. ft TDS Corepoint® Scientific Hazardous Location Manual Defrost Freezers, 17 cu. ft
Datasheet
More
Customers Who Bought This Also Bought