About this item
Get accurate water testing results with the Free Chlorine colorimeter. With a digital read out, there is no more struggling to determine colors visually. This free chlorine tester's compact design fits in your pocket or stores easily in your shop.
- Easier to use and more accurate than chemical test kits
- One-button operation makes getting your free chlorine results simple
- Great for water quality, environmental testing, and pools and spas
- For use with freshwater samples
- Easier to use and more accurate than chemical test kits
- Dedicated to a single parameter
- Small size, big convenience
- Accuracy: ±0.03 ppm, ±3% of reading
This Handheld Colorimeter is a simple, accurate and cost-effective way to measure free chlorine. Free chlorine is the active form of chlorine used for disinfection of drinking water, swimming pools and spas, and vegetable rinse water. Free chlorine in water is also harmful to a reverse osmosis membrane and needs to be monitored for removal from source water prior to being used. Designed as a more accurate alternative to chemical test kits, this free chlorine meter provides quick, accurate results in a few easy steps.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA470349-846
- CA470349-862
- Method:
- Adaptation of USEPA method 330.5, DPD method
- Ambient operating temperature:
- 0...50 °C (32...122 °F); RH max 95% non-condensing
- Resolution:
- 0.01 ppm