Specifications
- Description:Bluetooth® pH meter PEI body field app
- Operating temperature range:–5…+70 °C (23...158 °F) - LT
- pH range:0.00 - 12.00
Specifications
About this item
This wireless field pH meter has a durable PEI body and spherical sensing bulb. All readings are transmitted directly to your Apple or Android device.
- Take laboratory grade pH and temperature measurements using your smart phone or tablet
- Easily connect to the Hanna Lab App at the press of a button via Bluetooth wireless technology
- Group your data by time or notation
- Email it for storage or share it with friends or colleagues
- Great for aquariums, education, environmental, and more
- Connects to Hanna Lab App with a compatible Apple or Android Device via Bluetooth Smart Technology (Bluetooth 4.0)
- Up to five-point pH calibration with seven standard pH buffers available
- Calibration reminder
PEI body -PEI is suitable for a wide range of applications and excels in field measurements due to its durability. The shielding around the spherical glass tip minimizes breakage due to accidental bumping or dropping of the electrode. PEI is a high-quality plastic that is chemically resistant to many aggressive chemicals.
Real time data - Display of pH and temperature updated every second. Readings are automatically saved to a history file every hour, limited only by the available memory on the device.
GLP (Good Laboratory Practice) - Displays date and time of current calibration, offset, and average slope, along with calibrated buffers, mV values, temperature and slopes between each buffer.
Log and share data - Saved log files may be annotated with measurement-specific information. Data is easily shared via email in CSV or PDF format.
Help and Tutorials - Demo mode helps you explore features of the Hanna Lab App. General app information, HALO information, pH tutorial, maintenance tutorial, and contact information are available in help.