Thermo Scientific™ Barcoded Latch Racks for Matrix™ Storage Tubes
Supplier: Thermo Fisher Scientific
Certificates
About this item
Enhance sample storage at low temperatures, including vapor phase liquid nitrogen, with Thermo Scientific™ Matrix™ barcoded latch racks.
- Storage in robot-friendly ANSI-footprint rack
- Racks feature linear barcode, human readable ID and 2D barcode for ultimate sample tracking regardless of your barcode system
- Rack barcodes are compatible with our range of VisionMate readers
- Hinged lid rests in 'open' position on rack while conducting manual pipetting to prevent contamination of lid sitting on bench
- ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
- Latch rack lid can be lifted robotically for tube access with high-throughput automated applications
Barcoded latch racks feature a linear barcode, 2D barcode and human readable rack identification on three sides of the rack for the ultimate in sample tracking. Use in combination with Thermo Scientific 2D barcoded tubes, barcode readers, arrayers and capping equipment.
Certifications: Products are manufactured in ISO 7 class clean room environment according to ISO 14644-1:2015. Sterile products are sterilized by gamma irradiation according to ISO 11137; 10⁻³ Sterility Assurance Level (SAL). The vials and closures were tested and met the requirements of USP <88> Class VI plastics.
Customers Who Bought This Also Bought