Orders LinkContactUsLinkComponent
eZwest™ Lite Automated Western Device
eZwest™ Lite Automated Western Device
eZwest™ Lite automated Western device, 1 unit
Catalog # 76628-452
Supplier:   Genscript
Supplier #:  L00816
eZwest™ Lite Automated Western Device
Catalog # 76628-452
Supplier:  Genscript
eZwest™ Lite Automated Western Device
eZwest™ Lite Automated Western Device
eZwest™ Lite automated Western device, 1 unit
Price
$5,806.73
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
eZwest™ Lite is a fully automated western blotting device completing the entire process of blocking, antibody incubation, and washing steps. Automation of western blot incubation steps minimizes errors and variability typically associated with manual operations contributing to experimental reproducibility. eZwest frees researchers from the manual labor of traditional western blot.
  • Hands-free: one-time reagent loading followed by completely automated incubation and washing process
  • Easy-to-use: Set up in less than 10 minutes
  • Reliable: Comparable results to conventional manual method
  • Flexible: Fully customized procedures and solutions
  • Supports short and overnight incubation
  • Antibody recoverable: Recover primary antibody
  • Multi-Blot Processing: Incubate up to two membranes at once
The eZwest™ Lite device is compatible with the traditional reagents and protocols used for conventional western blotting. No proprietary reagents are required. Fully customizable steps and solutions make eZwest™ Lite a powerful tool fitting specific experiment needs for both routine and novel western blotting conditions.
GenScript’s eZwest™ Diluent kit is designed to be used in combination with the eZwest™ Lite Automated western device. The eZwest™ Diluent kit speeds up the overall western blotting process by shortening the incubation time for primary and secondary antibodies. Moreover, it can be used as a blocking reagent during the incubation, thus eliminating the need for a separate blocking step. The results obtained using the eZwest™ Diluent Kit are as sensitive as those obtained through a conventional western blotting protocol.
Specifications
  • Description:
    eZwest™ Lite automated Western device
  • Size:
    1 unit
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Membrane spacer, 50 piece
    Membrane spacer, 50 piece
    76683-240
    Log in for availability
    Each - $83.00
Product Information
  • Catalog #
    Availability
    Price
  • eZwest™ membrane cassette, 1 unit
    eZwest™ membrane cassette, 1 unit
    76683-242
    Log in for availability
    Each - $697.45
Product Information
  • Catalog #
    Availability
    Price
  • eZwest™ cleaning cassette, 1 unit
    eZwest™ cleaning cassette, 1 unit
    76683-246
    Log in for availability
    Each - $443.47
Product Information
  • Catalog #
    Availability
    Price
  • eZwest™ diluent kit, 1 unit
    eZwest™ diluent kit, 1 unit
    76683-238
    Log in for availability
    Each - $265.74