Fractional Distillation Set
Catalog #: 470343-788
Supplier: Wards
Specifications
- Description:Fractional distillation set
Specifications
About this item
Comprehensive four-piece set for accurate fractional distillation, including 250 ml distillation flask, Vigrex column, receiver adapter, and still head, all crafted from high-quality borosilicate glass.
- Fractional distillation process is the separation of a mixture into individual parts of fractions by heating it up
- Kit includes five pieces: 250 ml round bottom flask, Vigrex column, still head, receiver adaptor
- All joint sizes are 24/29
- All pieces are made of high quality, heavy duty borosilicate glass 3.3
- Perfect for any school or professional laboratory
Customers Who Bought This Also Bought