Simple Distillation Set
Catalog #: 470345-146
Supplier: Wards
Specifications
- Description:Simple distillation set
- Cat. no.:470345-146
Specifications
About this item
This four-piece set with 24/29 joints provides the necessary components for simple distillation processes, ensuring efficiency and reliability in laboratory work with its high-quality borosilicate glass construction.
- Simple Distillation set made of high quality Borosilicate glass
- All joint sizes are 24/29
- All pieces are made of high quality, heavy duty Borosilicate Glass 3.3
- Perfect for any school or professional laboratory
Set includes four pieces: 250 ml round bottom flask, still head, Liebig condenser, adaptor receiver.
Customers Who Bought This Also Bought