Rat fibroblast growth Factor 21( FGF-21) ELISA Kit
Catalog # 76706-304
Supplier: AFG Bioscience
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Assay Type:Sandwich
- Host:Rat
- Primary antibody reactivity:
- Target protein:Fibroblast growth factor 21
- Description:Rat fibroblast growth factor 21( FGF-21) ELISA kit
- Size:96 wells
- Environmentally Preferable:
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Assay Principle:Quantitative
- Shelf life:6 months (4 °C); 12 months (−20 °C)
- Detection range:3 - 200 pg/ml
- Storage temperature:2…8 °C
- Application:Cardiovascular, signal transduction
- Sample volume:50 - 100 μl
- Sensitivity:0.6 pg/ml
- Regulatory status:RUO
- Cat. no.:76706-304
Specifications
About this item
- Detection wavelength: 450 nm