ContactUsLinkComponent
Student Distillation Apparatus Kit
Student Distillation Apparatus Kit
Catalog # 470345-020
Supplier:  Wards
undefined
Student Distillation Apparatus Kit
Catalog # 470345-020
Supplier:  Wards
Supplier Number:  DIST1SET
Specifications
  • Description:
    Student distillation apparatus kit
  • Environmentally Preferable:
  • Cat. no.:
    470345-020
Specifications
About this item
Kit includes glassware components, stands, and borosilicate glass for student distillation experiments, providing a comprehensive and reliable setup for educational purposes.
  • Distillation apparatus kit
  • Necessary distillation glassware components and stands
  • Includes retort stand, clamp, tripod burner stand with wire gauze, rubber stoppers, Kjeldahl bulb, Liebig condenser, boiling flask and Erlenmeyer flask
  • Glassware made of high quality, borosilicate glass
  • Great for chemistry classrooms
This distillation apparatus kit includes stands and glassware components, perfect for jump starting your chemistry lab distillation. Set includes retort stand, clamp, tripod stand with wire gauze, rubber stoppers, Kjeldahl bulb, Liebig condenser, 500 ml boiling flask and 500 ml Erlenmeyer flask. Retort stand measures 26" tall, 8" wide and 8" long. Tripod burner stand measures 8" tall, 7" wide and long, and the wire gauze measures 5×5" and features a ceramic center. Liebig condenser measures 18" long. The 500 ml boiling flask, and the 500 ml Erlenmeyer flasks each measure 7" tall, and have bottom diameters of 4". Each flask neck has an inner diameter of 28 mm and an outer diameter measuring 33 mm. Autoclavable, with superior chemical durability. Low temperature gradient- can withstand high temperatures and thermal shocks. Eisco Borosilicate glass has the following heat tolerances: Strain point 515 °C, annealing point 565 °C, softening point 820 °C.