Starter kit for Simple Organic Chemistry
Catalog #: 470344-194
Supplier: Wards
Specifications
- Description:Starter kit for simple organic chemistry
Specifications
About this item
This 8-piece starter kit provides the essential components for simple organic chemistry experiments, ensuring beginners have the necessary tools made of high-quality materials for reliable performance and ease of use.
- The perfect starter set for organic chemistry labs, distillation, fractional distillation, separation, purification, and synthesis
- All pieces made of 3.3 borosilicate glass that is hand blown and polished for perfect fit
- Eisco E-Lab glass guarantee, any piece breaks during shipping will be replaced free of charge
- Metal ware stands not included in set
- Comes with Case and foam to hold all components
This set comprises 8 items of 14/23 ground joint. Beaker 25 ml vigreux column 100 mm liebig condenser 100 mm still head screw cap adapter receiver adapter pear shaped flask 25 ml thermometer −10 to +110 °C All pieces made with high quality borosilicate glass Eisco Borosilicate glass has the following heat tolerances: Strain point 515 °C, annealing point 565 °C, softening point 820 °C.
Customers Who Bought This Also Bought