Synthware Storage Vessel, #15 O-Ring Joint
Supplier: Kemtech America
Certificates
About this item
Storage vessel with a #15 O-ring joint on the sidearm and a high vacuum valve to control gas or liquid flow into the vessel.
- Heavy wall construction
- 33 expansion borosilicate glass
- Standard ground glass joints work with other brands
Supplied complete with one #116 Viton® O-ring, and one high vacuum valve.
Customers Who Bought This Also Bought