Synthware Gas Dispersion Tubes, Fritted, 90° Angle
Supplier: Kemtech America
Certificates
About this item
The 90° angle permits bubbles to be spread over a large area.
- Heavy wall construction
- 33 expansion borosilicate glass
- Standard ground glass joints work with other brands
For use with filtration techniques.
Customers Who Bought This Also Bought