Synthware Check Valve
Catalog #: 60004-922
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Connection type:8 mm hose connection
- Description:Check valve
- Supplier No.:V180008
Specifications
About this item
Gound seat for liquid seal.
- Heavy wall borosilicate construction
- All glass
- Ground seat to ensure liquid seal
Heavy wall check valve is designed to prevent gas or liquid backflow.
Customers Who Bought This Also Bought