Economy Calorimeter
Catalog #: 470349-676
Supplier: United Scientific Supplies
Specifications
- Description:Economy Calorimeter
- Cat. no.:470349-676
Specifications
About this item
Easily determine the caloric energy in food.
- Determine the caloric count of snack food
- Holds the food item safely
- Includes activity guide
The economy food calorimeter allows students to investigate the amount of energy released when food is burned. This involves using the burning energy of food to heat a vessel of water. The increase in temperature of the water represents the energy given off by the burning food. During the investigation students will learn how much energy is required to burn off some food items, and also how to read the caloric specification on food wrappers.
Customers Who Bought This Also Bought