Green Laser Pointer
Catalog #: 470343-196
Supplier: American Scientific
Specifications
- Description:Green laser pointer
Specifications
About this item
Bright green laser pointer, at a budget friendly price.
- Maximum power: <5 mW (class 3R/IIIa)
- Approximate wavelength: 652 nm
- Includes a teacher’s guide
- Includes 2 AA batteries in a reusable gift box
The color green is the most perceptible color to the human eye. This makes the green laser pointer ideal for applications where the beam must be observed. This laser pointer is perfect for diffraction wavelength comparisons to red lasers. Or, use it for demonstrating lenses, prisms, or mirrors, and in long-distance applications.
Customers Who Bought This Also Bought