About this item
The Blank Cryo Label kit is for glass or plastic tubes and vials exposed to −120 °C.
- Resistant to solvents such as DMSO, methanol, isopropyl alcohol and more
- Accomodates containers ranging in size from 0.5 to 50 ml
Both the 1 and 3'' core are compatible with thermal transfer printers. The 3'' core is compatible with the cab AXON2 and cab AXON1 printer/applicators.