ContactUsLinkComponent
Fragment Analyzer System CRISPR Kits
Fragment Analyzer System CRISPR Kits
Catalog #: CA76645-266
CRISPR DISCOVERY GEL KIT 1000 SAMPLES
Fragment Analyzer System CRISPR Kits
Catalog #: CA76645-266
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    CRISPR discovery gel kit, 1000 samples
  • Cat. no.:
    CA76645-266
Specifications
About this item
Screen CRISPR/Cas9 mutation events with heteroduplex cleavage assays.
  • Separate heteroduplex cleavage fragments - clear sizing and relative quantification data help you determine CRISPR gene editing success
  • Powerful data analysis - plugins for ProSize data analysis software automatically identify samples with CRISPR mutation events and provide important information such as percent mutated
  • ssDNA assessment - quality analysis of ssDNA and detection of incomplete dsDNA digestion aids in determining CRISPR/Cas homology directed repair (HDR) success
  • Low sample volume - only 2 µl of sample required minimizing amount of digestion reaction volume required
Efficient and accurate identification of CRISPR mutation events is a critical component of CRISPR/Cas9 workflows.  Heteroduplex cleavage assays offer a simple, accurate, and cost-effective method to identify CRISPR mutation events.
The CRISPR discovery gel kit for the Agilent fragment analyzer systems automates the screening of heteroduplex assays and provides fragment sizing and relative concentration. These important pieces of information are automatically processed by ProSize data analysis software to indicate successful editing events and other crucial information like percent mutated.
Caution: For research use only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought