Specifications
- Description:5200 fragment analyzer
- Cat. no.:CA76645-014
Specifications
About this item
Parallel capillary electrophoresis system used for nucleic acid QC in low throughput labs.
- Separate 12 samples in parallel in as little as 15 minutes
- Choose between three different capillary array lengths for the required blend of speed or resolution
- Minimize instrument preparation time with no daily array handling requirements and room-temperature stable reagents
- See clear results with separation resolution as good as 3 bp from fragments under 300 bp
- Improve lab efficiency by loading and programming samples while a separation is currently running
- Move samples up or down in queue to adjust run priorities
- Use quality metrics for RNA (RQN) and genomic DNA (GQN) to remove subjective quality assessments
- Kits provide a wide dynamic range covering two orders of magnitude; Achieve accurate molarity calculations with reliable smear analysis
- IQ/OQ services available for use in GMP environments
- Desktop PC supplied with a comprehensive 3-year warranty through HP
The 5200 Fragment Analyzer system is a parallel capillary electrophoresis instrument that improves lab efficiency allowing you to separate 12 samples in parallel in as little as 15 minutes. This system is designed to eliminate laboratory bottlenecks so you can focus on results.
The 5200 Fragment Analyzer system performs DNA QC and RNA QC for a broad range of samples including, gDNA, small RNA, cfDNA, large DNA fragments, and total RNA. The diversity of sample types these systems can separate make these instruments ideal for individualized workflows, including NGS library QC and CRISPR workflows.