Specifications
- Description:gDNA 165 kb analysis kit
- Cat. no.:CA76645-380
Specifications
About this item
Quality control of high molecular weight gDNA and long read NGS libraries.
- Reliable sizing - Accurate and precise sizing of HMW gDNA samples ranging from 1300 bp to 165 kb
- Reduced time to results - Optimized pulsed-field separation methods decrease QC analysis time and eliminates the need for overnight PFGE separations
- Quality metrics - Genomic Quality Number (GQN) quickly scores the integrity of HMW gDNA
- Ultra-sensitive detection - Input concentrations as low as 5 pg/µl
- Low sample volume - Only 2 µl required, conserving sample for downstream analysis
The genomic DNA 165 kb kit is used in the assessment of high molecular weight (HMW) genomic DNA quality control required for long-read NGS libraries. This kit is designed to run with an optimized pulsed-field separation method enabling accurate sizing of HMW gDNA through 165 kb.
Long-read NGS library preparations require several gDNA quality control steps. Compared to standard 16-hour PFGE protocols, the Genomic DNA 165 kb kit produces results in as little as 1.5 hours. Replacing traditional PFGE with the Genomic DNA 165 kb kit eliminates days from the tedious library workflow.