About this item
These flammable storage refrigerators offer premium cooling and protection for flammable substances.
- Microprocessor temperature controller
- Temperature alarms
- Keyed door locks
- Probe access
- Natural hydrocarbon refrigerants
- ENERGY STAR certified*
These flammable storage refrigerators meet the requirements of NFPA 45 and NFPA 30 for the cold storage of flammable and combustible materials in unclassified laboratory locations. They utilize non-sparking and/or ATEX rated components to ensure an intrinsically safe interior for flammable materials in sealed containers. These units feature temperature alarms, keyed door locks, interior shelving, probe access, and microprocessor temperature control.
The Performance Series includes premium additions such as state-of-the-art touchscreen controllers, advanced alarms and data logging capabilities and unparalleled temperature control. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
*Select models ENERGY STAR certified: 76644-724
Specifications
- Catalog No:
- 76644-728
- 76644-724
- 76644-720
- Model:
- 76644-728
- 76644-724
- 76644-720
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Defrost:
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Shelves/drawers:
- 5 adjustable shelves (4 full, 1 half shelf)
- 5 adjustable shelves (4 full, 1 half shelf)
- 5 adjustable shelves (4 full, 1 half shelf)
- Door type:
- 1 solid door, right hinged
- 1 solid door, right hinged
- 1 solid door, right hinged
- Weight:
- 114 kg (252 lbs.)
- 110 kg (242 lbs.)
- 87 kg (192 lbs.)
- Environmental certification:
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants