Mobile Phase Reservoir Bottle, MicroSolv
Catalog #: 97057-216
Supplier: MicroSolv Technology Corp
Specifications
- Capacity:1 L
- Description:Mobile phase reservoir bottle, glass, clear, standard taper top, 29/32 neck
- Cat. no.:97057-216
- Supplier No.:58801-01
Specifications
About this item
This borosilicate glass bottle is perfect for HPLC mobile phase reservoirs with its small opening and large capacity. Graduations on the outside of the bottle help you determine the Solvent use and prevent running dry.
This Bottle requires a S/T (Standard Taper) 29/32 joint for the mouth and will produce a tight seal. These bottles are often used on older HPLC Instruments such as the Agilent 1090.
Customers Who Bought This Also Bought