HPLC Mobile Phase Storage Bottle, MicroSolv
Catalog #: 75834-186
Supplier: MicroSolv Technology Corp
Specifications
- Capacity:100 ml
- Description:Bottles, HPLC mobile phase storage, GL45
- Cat. no.:75834-186
- Supplier No.:58807-100-A-CS
Specifications
About this item
These amber borosilicate glass bottles have a 100 ml 'Total Fill Capacity' and are perfect as HPLC Mobile Phase Storage Bottles. The small openings and enamel graduations and marking patches makes them a convenient way to store concentrated Mobile Phase or its Ingredients.
Blue, solid screw caps without liners and polypropylene drip rings are included. This bottle is not 'safety coated'.
Customers Who Bought This Also Bought