To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Calibration:Automatic, 1 or 2 points choose between 2 sets of buffers (standard: 4.01; 7.01; 10.01)
Automatic, one-point at 1413 μS/cm (Conductivity)
Automatic, one-point at 1382 ppm (CONV=0.5) or 1500 ppm (CONV=0.7) (TDS) - Description:Portable waterproof pH/EC/TDS meter (low range)
- Conductivity accuracy:±2% F.S. at 25 °C/77 °F
- Conductivity range:0 - 3999 μS/cm
- Conductivity resolution:1 μS/cm
- TDS accuracy:±2% F.S. at 25 °C/77 °F
- TDS range:0 - 2000 ppm
- TDS resolution:1 ppm (mg/L)
- pH accuracy:±0.02 pH/±0.1 pH at 25 °C/77 °F
- pH range:–2...+16
- pH resolution:0.01 pH/0.1 pH
Specifications
About this item
Designed to bring simplicity to environmental testing, this pH/EC/TDS Meter is designed specifically for use in manufacturing and environmental testing protocols. The specialized electrode combines 3 sensors in a single rugged probe body featuring a polypropylene body, integrated temperature sensor, and cloth junction.
- 3-in-1 probe measures pH, EC/TDS, and temperature
- Comes with all the necessary solutions and batteries - everything you need to get started measuring right away
- Perfect for water quality testing
- Probe condition - an on-screen indicator provides visual confirmation that your probe is working at its best
This meter is designed to withstand the knocks, drops, and spills of real life, the new IP 67 body ensures top performance in any environment. These meters are protected against dust and water intrusion from any direction. Also has a portable design, Made to fit comfortably in your hand for on-the-go testing, the multilevel LCD display provides at-a-glance readings of pH and temperature from any angle and dual button operation streamlines the testing process for any user. The probe connection is a quick Din connection. A Quick Connect DIN connector makes attaching and removing the probe simple and easy. The rubber coating protects the cable and provides a water-tight seal without twisting.
This pH/EC/TDS meter features a replaceable HI12883 probe that has been carefully designed specifically for pH measurements encountered in manufacturing and environmental testing protocols. The polypropylene body of the HI12883 houses all three sensors within a sturdy, single body design. The plastic body protects the electrode from damage, is easy to clean, and is chemically resistant to many aggressive chemicals. The pH sensor in the probe is a single junction electrode with gelled electrolyte and a retractable cloth junction. When readings appear sluggish, simply pull out some of the cloth material from out from its position t renew the junction with an uncontaminated, fresh surface. The EC/TDS portion is a two-ring probe that comes with the HI991300, offering an accurate solution for low range EC and TDS readings. An integrated temperature probe ensures fast, accurately compensated readings even during sudden temperature fluctuations.
Low battery indicator - the included batteries provide 1400 hours of continuous use. When the battery level is below 10%, a warning symbol will blink to indicate low battery condition. When low battery power may affect your results, the meter will shut down to avoid erroneous readings caused by low battery level.