About this item
Tygon S3™ tubing is phthalate-free and is made from a NON-DEHP/bio-based plasticizer.
- Outstanding chemical resistance
- Consistent for reproducible results
- Easily slips over fittings and grips securely for simple lab set-ups
- Standard sizes are able to hold a full vacuum
This sustainable, flexible, long lasting, crack-resistant tubing continues to provide the same great performance.
The sterilizing methods for this tubing are conventional autoclaving (steam 30 minutes at 15 psi, 121 °C) and ethylene oxide.
Tygon S3™ Formulation E-3603 tubing is non-oxidizing, non-contaminating, and less permeable than rubber tubing. The glassy smooth inner bore helps prevent build-up and facilitates cleaning. Coils are also marked at 30.5 cm (1') intervals for easy measuring. It is specially formulated for resistance to flex-fatigue and abrasion. Generally used as tubing in instrumentation connection, venting, draining, and other laboratory applications, Tygon S3™ Formulation E-3603 tubing offers superior life, minimizing labor and the expense of replacement.
Specifications
- Catalog No:
- 89403-854
- 89403-876
- 89403-856
- 89403-878
- 89403-900
- 89403-872
- 89403-894
- 89403-896
- 89403-864
- 89403-886
- 89403-882
- 89403-848
- 89403-850
- 89403-868
- 89403-890
- Model:
- 89403-854
- 89403-876
- 89403-856
- 89403-878
- 89403-900
- 89403-872
- 89403-894
- 89403-896
- 89403-864
- 89403-886
- 89403-882
- 89403-848
- 89403-850
- 89403-868
- 89403-890
- Durometer hardness:
- Shore A, 56
- Shore A, 56
- Max. vacuum:
- 29.9" Hg @ 73 °F
- 13.0" Hg @ 73 °F
- 9.0" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 9.0" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 5.0" Hg @ 73 °F
- 21.0" Hg @ 73 °F
- 20.0" Hg @ 73 °F
- 21.0" Hg @ 73 °F
- 9.0" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- 29.9" Hg @ 73 °F
- Working pressure:
- 36.0 psi
- 17.0 psi
- 14.0 psi
- 30.0 psi
- 14.0 psi
- 36.0 psi
- 12.0 psi
- 20.0 psi
- 20.0 psi
- 20.0 psi
- 14.0 psi
- 36.0 psi
- 25.0 psi
- 28.0 psi
- 25.0 psi
Specifications
Related Information
Certificates
Search by Lot Number89403-848_24539411 89403-848_24758157 89403-848_25004458 89403-854_25191033 89403-854_31356263