ADC® EKG Calipers
Catalog #: 470349-496
Supplier: AMERICAN DIAGNOSTIC CORP CP
Specifications
- Description:EKG caliper
- Cat. no.:470349-496
Specifications
About this item
Made with hardened steel arms and points for accurate measurement of tracing intervals.
- Point protector has built-in 40 mm metric scale measures 4¾"
- Optional retail packaging (see photos) is available for a slight upcharge
- Add a ‘Q’ to the item number when ordering
Designed to measure EKG waveforms.
Customers Who Bought This Also Bought