Adenosine 5'-monophosphate disodium salt (AMP disodium salt)
Catalog #: CA80050-108
Supplier: MilliporeSigma
CAS Number: 4578-31-8
Specifications
- Packaging:Plastic ampoule
- Size:5 g
- Cat. no.:CA80050-108
- Supplier No.:118110-5GM
Specifications
Specification Test Results
- Adenosine 5'-monophosphate disodium salt (AMP disodium salt):
Specification Test Results
About this item
Synonyms: 5′-AMP-Na2
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 391.2 g/mol
:Adenosine 5'-monophosphate disodium salt (AMP disodium salt)
Formula:C₁₀H₁₂N₅Na₂O₇P
MW:391.2 g/mol
MDL Number:MFCD00065023
CAS Number:4578-31-8
Customers Who Bought This Also Bought