To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Mesothelin
- Clonality:Monoclonal
- Clone:YP158
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Conjugation:Unconjugated
- Size:6 ml
- Cat. no.:CA76634-884
Specifications
About this item
Mesothelin is a glycocylphosphotidylianositol-linked cell-surface glycoprotein, which is present on the surface of normal mesothelium and is overexpressed in many patients with epithelial ovarian cancer and malignant mesotheliomas.