To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Myeloperoxidase
- Clonality:Monoclonal
- Clone:59A5
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Conjugation:Unconjugated
- Size:6 ml
- Cat. no.:CA76634-528
Specifications
About this item
Myeloperoxidase metabolizes most of the hydrogen peroxide generated by activated phagocytes. This antibody stains strongly with the cytoplasm of myeloid cells. The antibody detects myeloperoxidase found in myeloid cells of acute myeloid leukemia. The myeloperoxidase gene has been localized to chromosome 17.