To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD10
- Clonality:Monoclonal
- Clone:56C6
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Conjugation:Unconjugated
- Size:6 ml
- Cat. no.:CA76634-356
Specifications
About this item
The common acute lymphoblastic leukemia antigen (CALLA/CD10) is a single-pass type II transmembrane metallo-endopeptidase that cleaves and inactivates a variety of peptide growth factors important for signal transduction including the enkephalins, bombesin and substance P. CD10 is expressed by a number of hematopoietic cells such as immature T and B cells, B cells of the germinal centers of lymphoid follicles and granulocytes. It also reacts with a variety of non-hematopoietic cells, including epithelial cells in GI tract and kidney tubular. In liver, the bile canaliculi shows a moderate to strong staining. CD10 has been used for the identification and classification of certain types of malignant lymphoma and leukemia. CD10 is expressed in a high percentage of cases of acute lymphoblastic leukemia, follicular lymphoma, Burkitt's lymphoma, some hematopoietic tumors, and chronic myelogenous leukemias in lymphoid blast crisis. It is also known to be a marker of endometrial stromal cells. It is helpful in differentiating endometrial stromal sarcoma (ESS) from uterine cellular leiomyoma (UCL) and uterine leiomyosarcoma (ULS).