To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD99
- Clonality:Monoclonal
- Clone:HO36-1.1
- Host:Mouse
- Isotype:IgM
- Reactivity:Human
- Conjugation:Unconjugated
- Size:6 ml
- Cat. no.:CA76634-354
Specifications
About this item
This antibody is specific to a sialoglycoprotein of 27 to 32 kDa known as CD99 or MIC2 gene product. CD99 is expressed on the cell membrane of some lymphocytes, cortical thymocytes and granulosa cells of the ovary, sertoli cells of testis, pancreatic islet cells and some endothelial cells. Mature granulocytes express very little or no CD99.