About this item
This antibody is specific to a 45 kDa protein, which is identified as MyoD1. This antibody is specific to an epitope of amino acid 3-56 in the N-terminus of mouse MyoD1. This antibody does not react with myogenin, Myf5 or Myf6. MyoD1 stains the nuclei of myoblasts in developing muscle tissues. MyoD1 is not detected in normal adult tissue but is expressed strongly in the tumor cell nuclei of rhabdomyosarcomas.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA76633-582
- CA76633-580
- CA76633-578
- Antigen name:
- MyoD1
- MyoD1
- MyoD1
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- 5.2F
- 5.2F
- 5.2F
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG2a
- IgG2a
- IgG2a