To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD3
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Conjugation:Unconjugated
- Size:6 ml
- Cat. no.:CA76634-600
Specifications
About this item
The clone F7.2.38 is specific for the non-glycosylated epsilon chain of the human CD3 molecule. Clone F7.2.38 recognizes T cells in the thymus, bone marrow, peripheral lymphoid tissue and blood and is a pan T cell marker. The CD3 molecule consists of five different polypeptide chains with molecular weights ranging from 16 to 28 kDa. The CD3 antigen is first detected in early thymocytes, and its appearance probably represents one of the earliest signs of commitment to the T cell lineage.