To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:SALL4
- Clonality:Monoclonal
- Clone:6E3
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Conjugation:Unconjugated
- Size:6 ml
- Cat. no.:CA76634-588
Specifications
About this item
Sal-like protein 4 (SALL4) is a zinc-finger transcription factor that serves as a master regulator of embryonic pluripotency and is involved in processes associated with stem cell activities. SALL4 expression in germ cells makes it a useful marker for germ cell tumors such as seminoma, embryonal carcinoma, yolk sac tumors and teratomas. SALL4 expression is also seen in the spermatogonia of normal testis.