About this item
Wilms tumor-1 (WT-1) is a gene involved in the induction of Wilms tumor, a pediatric renal malignancy. Wilms tumor is associated with mutations of WT1, a zinc-finger transcription factor that is essential for the development of the metanephric kidney and the urogenital system. The WT1 gene is normally expressed in fetal kidney and mesothelium, and its expression has been suggested as a marker for Wilms tumor and mesothelioma.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA76633-914
- CA76633-910
- CA76633-912
- Antigen name:
- Wilms tumor 1 protein (WT1)
- Wilms tumor 1 protein (WT1)
- Wilms tumor 1 protein (WT1)
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- 6F-H2
- 6F-H2
- 6F-H2
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- IgG1