To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:HLA-DR (ß chain of DP DQ and DR)
- Clonality:Monoclonal
- Clone:CR3/43
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Conjugation:Unconjugated
- Size:0.1 ml
- Cat. no.:CA76633-218
Specifications
About this item
This antibody reacts with the β chain of all products of the gene sub regions DP, DQ, and DR. It stains B cells, interdigitating reticulum cells, Langerhans cells and many macrophages. This antibody does not react with normal T cell and polymorphs, but stains activated T cells.