About this item
This antibody reacts with an 80 kDa protein known as hCdc47. hCdc47 is a member of the MCM family, which allows DNA to replicate once per cell cycle. Expression of Cdc47 protein has been found in the nuclei of proliferative components of normal lymph nodes, bone marrow, epidermis and mucosa. Malignant tumors from several organs contain more nuclear hCdc47-positive cells than their normal counterparts.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA76633-714
- CA76633-710
- CA76633-712
- Antigen name:
- CDC47
- CDC47
- CDC47
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- 47DC141
- 47DC141
- 47DC141
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- IgG1