To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:TdT
- Clonality:Monoclonal
- Clone:SEN 28
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Conjugation:Unconjugated
- Size:6 ml
- Cat. no.:CA76634-344
Specifications
About this item
This antibody stains a 60 kDa peptide in human lymphoblasts. TdT is present in the cells during immunoglobulin gene rearrangement and plays an important role in lymphocyte maturation. TdT marks both T and B immature lymphocytes normally found in thymus and bone marrow.