About this item
This antibody is specific to a cluster of proteins between 19-22 kDa (reduced). VEGF is a homodimeric disulfide-linked glycoprotein involved in angiogenesis, which promotes tumor progression and metastasis. It exhibits potent mitogenic and permeability-inducing properties specific for the vascular endothelium.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA76634-432
- CA76633-640
- CA76633-642
- CA76633-638
- Antigen name:
- Vascular endothelial growth factor (VEGF)
- Vascular endothelial growth factor (VEGF)
- Vascular endothelial growth factor (VEGF)
- Vascular endothelial growth factor (VEGF)
- Antibody type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- VG1
- VG1
- VG1
- VG1
- Reactivity:
- Human
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa