About this item
This antibody is specific to 53 kDa protein, expressed on activated platelets. CD63 is a lysosomal membrane glycoprotein that is translocated to plasma membrane after platelet activation. It is also expressed on granulocytes, B cells and T cells.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA76633-628
- CA76633-630
- CA76633-626
- Antigen name:
- CD63
- CD63
- CD63
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- NK-1/C3 (same as MX 493129.5)
- NK-1/C3 (same as MX 493129.5)
- NK-1/C3 (same as MX 493129.5)
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa