To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Alpha fetoprotein
- Clonality:Monoclonal
- Clone:C3
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Conjugation:Unconjugated
- Size:6 ml
- Cat. no.:CA76634-314
Specifications
About this item
Alpha fetoprotein (AFP) is a single chain glycoprotein of 70 kDa that is normally expressed in the fetal liver, gastrointestinal tract and yolk sac. This antibody recognizes an epitope of α-fetoprotein present in human and several other species.