Bacto™ Casamino Acids, Technical, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Bacto Casamino Acids, technical, is an animal origin (AO) acid hydrolysis of casein. In contrast to the Bacto Casamino Acids, this supplement has been processed less and, as a result, has higher sodium chloride and iron content.
- An animal origin (AO) acid hydrolysis of casein
- Less processing results in higher sodium chloride and iron content
- Used in the production of animal health vaccines
- Supports the growth of Bordetella, Haemophilus, and Vibrio species
Bacto Casamino Acids, Technical provides similar benefits to Bacto Casamino Acids, for applications requiring a less refined hydrolysate.
Certifications: AOAC, BAM, COMPF, SMWW, USDA, USP, ISO/DIS, 11014.
Customers Who Bought This Also Bought