Accessories for Multidrop® Combi Microplate Dispensers, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
The comprehensive line of Thermo Scientific™ Multidrop™ dispensing cassettes offers optimal solutions for dispensing a wide range of volumes and reagents for excellent performance and results.
- Designed for use with Multidrop Combi SMART+ dispensers
- Equipped with radio frequency identification (RFID) tags to store lifetime dispensing data and display remaining usage time
- Available in standard and small tube versions
- Distinguished from normal cassettes by antenna tags
The Multidrop Combi+ Reagent Dispenser offers ultimate flexibility for reagent dispensing. Multidrop Combi SMART+ dispensers have an advanced built-in cassette lifetime tracking system.
Related Information
Multidrop Combi Dispenser Multidrop Combi Reagent Dispensers
Additional Product Details
Multidrop Dispensing Cassettes
Application Notes
Customers Who Bought This Also Bought