ContactUsLinkComponent
Smiths Medical® Large-Bore Stopcock Fittings, Male Luer Lock
Smiths Medical® Fitting, Polycarbonate, Three-Way, Large-Bore Stopcock with Male Luer Lock; 1000/PK
Catalog #: MFLX30600-23BLK
Smiths Medical® Fitting, Polycarbonate, Three-Way, Large-Bore Stopcock with Male Luer Lock; 1000/PK
Catalog #: MFLX30600-23BLK
Specifications
  • Description:
    Fitting, Polycarbonate, Three-Way, Large-Bore Stopcock with Male Luer Lock; 1000/PK
  • Wetted materials:
    Polycarbonate
  • Warranty:
    Out of box
  • Cat. no.:
    MFLX30600-23BLK
  • Flow Pattern:
    Three-Way
  • Inlet Port:
    Female luer
  • Outlet Port:
    Male luer lock
Specifications
About this item
Enhance fluid flow with a larger bore than standard luer stopcocks.
  • Three-way flow pattern
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Large-bore adapters are perfect for handling higher flow volumes in process while using connections similar to luer systems.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
  • Description:
    Fitting, Polycarbonate, Three-Way, Large-Bore Stopcock with Male Luer Lock; 1000/PK
  • Wetted materials:
    Polycarbonate
  • Warranty:
    Out of box
  • Cat. no.:
    MFLX30600-23BLK
  • Flow Pattern:
    Three-Way
  • Inlet Port:
    Female luer
  • Outlet Port:
    Male luer lock
Customers Who Bought This Also Bought