Specifications
- Sterility:Sterile
- Description:Klerwipe™ Low residue peroxide wipes
- Cat. no.:76637-530
- Packaging:Pouch
Specifications
About this item
Klerwipe™ Low residue peroxide pouch wipes are ideal for routine or rotational use when cleaning and residue removal is required.
- Sterile, knitted 100% polyester wipes
- Double bagged in a resealable pouch
- Filled, sealed, packed, and double bagged in an ISO Class 5 cleanroom
- Ultrasonically sealed edge for reduced fiber contamination
Klerwipe™ Low residue peroxide pouch wipes are non-corrosive and suitable for use on all types of surfaces in cleanrooms and controlled environments, isolators, LAFs, equipment exteriors, and production vessels. These wipes can be used to clean or remove residues without the need to wipe down with alcohol or water afterwards. These wipes are highly absorbent and remain wet throughout use, ensuring a consistent application of Low Residue Peroxide to the entire surface area. The use of wiping can increase cleaning consistency and the quality of the final product.
Ready-to-use, quick and easy to apply, Klerwipe™ Low residue peroxide pouch wipes are impregnated with Klercide™ Low residue peroxide, a non-corrosive formulation that leaves minimal residues. These non-corrosive wipes are powerful and effective in a wide range of cleanroom applications. The wipes are packaged in a resealable pouch. Each pouch is designed to ensure it is easy to open and reseal during use by gloved operators. Each resealable pouch contains 10 wipes, which helps reduce waste.