Innovating Science® Ocean Acidification
Catalog #: 470348-620
Supplier: Ward's Science
Specifications
- Description:Ocean acidification kit
- Includes:Instructions and SDS
- No. of Uses:15
Specifications
About this item
Learn about the carbon cycle and how it relates to ocean acidification.
- Three different activities for student investigations
- Determine the cause of ocean acidification and the effects it can have on the ecosystem and marine life
Changes to the carbon cycle and climate due to human activities have had detrimental effects on the environment. One example of this is known as ocean acidification, which is the changing of the ocean's chemistry due to increased carbon dioxide in the atmosphere and ocean. In this activity, students will learn about the carbon cycle and how it relates to ocean acidification. Through three different activities, students will investigate the cause of ocean acidification, and the effects it can have on the ecosystem and marine life. Kit contains enough materials for 15 groups of students. Teacher's manual and student study guide copymasters are included.
Customers Who Bought This Also Bought