Specifications
- Description:Bioquell ProteQ Bio-decontamination system
Specifications
About this item
The Bioquell ProteQ offers a modular approach to room and zone bio-decontamination. This latest generation room bio-decontamination system from Ecolab offers enhanced distribution capabilities, networking options, wireless communication technology and advanced aeration capabilities. It is an ideal solution for microbial contamination elimination from small labs to the large production areas.
- Mobile hydrogen peroxide vapor generator for chambers or rooms
- Advanced distribution and aeration capabilities with wireless communication provide faster setup and cycle times
- 6-log sporicidal kill on exposed surfaces
- Peroxide traceability and wastage minimization through the two-bottle module system
- A single system is ideal for rooms less than 400 m³
- Multiple systems can be used for spaces greater than 400 m³
- Utilizes EPA registered sterilant 35% w/w aqueous hydrogen peroxide solution (EPA registration number 72372-1-86703)
- Designed with RFID label technology for traceability and auditing
- 21 CFR Part 11 audit trail compliance package
- Integration testing for BMS and more complex systems
- Cycle recording (thermal printer and solutions for real time cycle data, including electronic records in pdf format)
- Supports GMP compliance
The Bioquell ProteQ offers a modular approach to room and zone bio-decontamination. This latest generation room bio-decontamination system from Ecolab offers enhanced distribution capabilities, networking options, wireless communication technology and advanced aeration capabilities.
Whether for routine or emergency remediation, make automated room and zone bio-decontamination technology a part of your contamination control with the Bioquell ProteQ. From small labs to large bioprocessing areas, the system boasts Ecolab’s Bioquell hydrogen peroxide vapor bio-decontamination technology which utilizes an EPA registered sterilant in Bioquell hydrogen peroxide sterilant (EPA registration number (72372-1-86703).
The Bioquell ProteQ offers built-in aeration, but cycles can be optimized further with optional wirelessly controlled aeration units stored directly in the system’s frame. Bio-decontaminate exposed surfaces in a single cycle easily for a 6-log sporicidal kill.
The body of the unit contains the hydrogen peroxide dispersion and distribution system for rooms up to 400 m³ (depending on configuration, load and environmental conditions). It contains a dual bottle module with RFID reading for large volume bio-decontamination and key data collection. Built-in aeration capability comes as a standard feature in every Bioquell ProteQ.
Up to three Bioquell ProteQ systems can be run simultaneously from a single master controller, making larger spaces simple to bio-decontaminate.