Specifications
- Description:Bioquell SeQure Bio-Decontamination System
- Cat. no.:76635-466
Specifications
About this item
The Bioquell SeQure is a fixed and compact wall-mounted bio-decontamination system. The technology utilizes Bioquell Hydrogen Peroxide Sterilant to kill microorganisms on exposed surfaces in spaces <150m3 . It offers a small footprint with fast installation and customized validated cycles performed by Ecolab validation experts on-site.
- Fixed hydrogen peroxide vapor generator for chambers or rooms
- Create validated cycles for 6-log sporicidal kill on exposed surfaces utilizing Bioquell Hydrogen Peroxide Sterilant (EPA Registration Number 72372-1-86703)
- Fast to install and validate (<1 week)
- Bottle module with peroxide traceability and wastage minimization through the two-bottle module system
- Ideal for pass-throughs, material airlocks, small enclosures and rooms less than 150m3 (5297 ft3)
- Designed with RFID label technology for traceability and auditing
- 21 CFR Part 11 audit trail compliance package
- Integration testing for BMS and more complex systems
- Cycle recording (thermal printer and solutions for real time cycle data, including electronic records in pdf format)
- Supports GMP compliance
The Bioquell SeQure is a fixed and compact wall-mounted bio-decontamination system. The technology utilizes Bioquell Hydrogen Peroxide Sterilant (EPA Registration Number 72372-1-86703) to kill microorganisms on exposed surfaces in spaces <150m3. It offers a small footprint with fast installation and customized validated cycles performed by Ecolab validation experts on-site.
The wall-mounted system is ideal for pass-throughs, material airlocks, and more. The Bioquell SeQure system features automatic cycle prediction technology for fast validation. Whether installed in a brand-new facility or retrofitted into an existing workspace, the Bioquell SeQure can be installed and validated in facilities in less than one week. The slim design and digital, user-friendly interface make the Bioquell SeQure ideal for use in many rooms and chambers. Its minimal footprint is enabled by wall-mounting the components of the system inside or outside of the workspace.