Specifications
- Description:Bioquell L-4, Hydrogen Peroxide Vapor Bio-Decontamination Generator
- Cat. no.:76635-462
Specifications
About this item
The Bioquell L-4 is a mobile Hydrogen Peroxide Vapor bio-decontamination generator that can be connected to enclosures, equipment, isolators and more in your facility or, with additional accessories, be used for room/zone bio-decontamination.
- Create validated cycles for 6-log sporicidal kill on exposed surfaces utilizing Bioquell Hydrogen Peroxide Sterilant (EPA Registration Number 72372-1-86703)
- Adaptable to various equipment types including Biosafety cabinets, closed systems and isolators, cage and rack washers, pass-throughs, material airlocks, freeze dryers and more
- Optional room bio-decontamination through distribution head for areas smaller than 350m3 (12360 ft3)
- Utilizes EPA registered sterilant 35% w/w aqueous hydrogen peroxide solution
- Designed with RFID label technology for traceability and auditing
- 21 CFR Part 11 audit trail compliance package
- Integration testing for BMS and more complex systems
- Cycle recording (thermal printer and solutions for real time cycle data, including electronic records in pdf format)
- Remote start stop integration and Automation
- Supports GMP compliance
The Bioquell L-4 is a mobile Hydrogen Peroxide Vapor generator that can be connected to enclosures, equipment, isolators and more in your facility or, with additional accessories, be used for room/zone bio-decontamination.
Disinfect equipment or small enclosure systems, open spaces or rooms, and more using the multipurpose Bioquell L-4. Bio-decontamination can be achieved through quick and direct connections to equipment and ports, open spaces, or for custom engineering solutions. Bio-decontaminate isolators, BSCs, lyophilizers, pass-throughs, rack washers and more when connected to the Bioquell L-4.
Room bio-decontamination options further showcase the system’s versatility. Use the Bioquell L-4 inside a room by attaching a distribution head, or outside of a zone with connection solutions to meet your needs.
Ecolab’s Bioquell Hydrogen Peroxide Sterilant (EPA Registration Number: 72372-1-86703) creates a 6-log sporicidal kill on exposed surfaces when used with Ecolab's Bioquell bio-decontamination equipment. You can operate Bioquell systems at ambient room temperature and relative humidity, without the need to attain significantly reduced humidity commonly associated with other hydrogen peroxide bio-decontamination systems.