Microcentrifuge Tubes
Catalog #: 470173-924
Supplier: Dynalon
Specifications
- Capacity:1.5 ml
- Description:Microcentrifuge tubes
- Material:PP
- Supplier No.:399325
Specifications
About this item
These microcentrifuge tubes are made of polypropylene. The attached lid provides a tight, leakproof seal.
- Leak tested to 20000 RCF for 20 minutes at 20 °C with an aqueous solution
- Lid membrane is uniformly thick to ensure trouble free piercing
- With frosted writing area and graduations
Customers Who Bought This Also Bought